Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Neurod4
Entrez Gene ID
288821
Gene Name
neuronal differentiation 4
Gene Aliases
-
Chromosome Location
Chr.7: 9699964 - 9712075 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105942.1 NP_001099412.1 1 - 2 71 82

View Full Product Details