The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108650.2 | NP_001102120.1 | 3 - 4 | 377 | 63 |
XM_008763394.1 | XP_008761616.1 | 3 - 4 | 1108 | 63 | |
XM_008763395.1 | XP_008761617.1 | 3 - 4 | 1108 | 63 | |
GenBank mRNA | BC160872.1 | - | 3 - 4 | 377 | 63 |
FQ211769.1 | - | 3 - 4 | 472 | 63 |