Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco5a1
Entrez Gene ID
312907
Gene Name
solute carrier organic anion transporter family, member 5A1
Gene Aliases
-
Chromosome Location
Chr.5: 5866550 - 5994748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107898.1 NP_001101368.1 1 - 2 1560 87
XM_006237743.2 XP_006237805.1 2 - 3 1651 87
XM_008763495.1 XP_008761717.1 2 - 3 1957 87

View Full Product Details