The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107898.1 | NP_001101368.1 | 8 - 9 | 2712 | 88 |
XM_006237743.2 | XP_006237805.1 | 9 - 10 | 2827 | 88 | |
XM_008763495.1 | XP_008761717.1 | 9 - 10 | 3133 | 88 |