Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ube2j2
Entrez Gene ID
298689
Gene Name
ubiquitin-conjugating enzyme E2, J2
Gene Aliases
-
Chromosome Location
Chr.5: 173372659 - 173387084 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007655.1 NP_001007656.1 5 - 6 767 83
XM_006239554.2 XP_006239616.1 5 - 6 619 83
XM_006239555.2 XP_006239617.1 6 - 7 954 83
XM_006239557.2 XP_006239619.1 4 - 5 614 83
XM_006239558.2 XP_006239620.1 4 - 5 456 83
XM_008764352.1 XP_008762574.1 5 - 6 777 83
XM_008764353.1 XP_008762575.1 5 - 6 1847 83
XM_008764354.1 XP_008762576.1 6 - 7 958 83
XM_008764355.1 XP_008762577.1 6 - 7 1903 83
GenBank mRNA BC079115.1 - 5 - 6 767 83

View Full Product Details