Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Heyl
Entrez Gene ID
313575
Gene Name
hes-related family bHLH transcription factor with YRPW motif-like
Gene Aliases
-
Chromosome Location
Chr.5: 140923914 - 140940905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107977.1 NP_001101447.1 3 - 4 544 63

View Full Product Details