The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009422.1 | NP_001009422.1 | 4 - 5 | 619 | 94 |
GenBank mRNA | BC087643.1 | - | 4 - 5 | 619 | 94 |
FQ219476.1 | - | 4 - 5 | 671 | 94 |