The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134619.1 | NP_001128091.1 | 6 - 7 | 3125 | 155 |
XM_006235428.2 | XP_006235490.1 | 7 - 8 | 3202 | 155 | |
XM_008762375.1 | XP_008760597.1 | 7 - 8 | 3965 | 155 | |
GenBank mRNA | FQ216244.1 | - | 6 - 7 | 838 | 155 |