Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alkbh5
Entrez Gene ID
303193
Gene Name
AlkB family member 5, RNA demethylase
Gene Aliases
RGD1309496
Chromosome Location
Chr.10: 46906314 - 46927759 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191643.1 NP_001178572.1 2 - 3 856 60

View Full Product Details