Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpo
Entrez Gene ID
287610
Gene Name
lactoperoxidase
Gene Aliases
-
Chromosome Location
Chr.10: 75100385 - 75120320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105829.1 NP_001099299.1 6 - 7 920 114
XM_006247085.2 XP_006247147.1 6 - 7 993 114

View Full Product Details