Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kremen2
Entrez Gene ID
287102
Gene Name
kringle containing transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.10: 13075950 - 13081136 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105767.1 NP_001099237.1 4 - 5 495 63
XM_008767550.1 XP_008765772.1 3 - 4 718 63
XM_008767551.1 XP_008765773.1 3 - 4 718 63
XM_008767552.1 XP_008765774.1 3 - 4 718 63

View Full Product Details