Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl3
Entrez Gene ID
306129
Gene Name
F-box and leucine-rich repeat protein 3
Gene Aliases
Fbxl3a
Chromosome Location
Chr.15: 93647307 - 93667395 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252442.2 XP_006252504.1 1 - 2 368 65

View Full Product Details