Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifb3
Entrez Gene ID
499924
Gene Name
BPI fold containing family B, member 3
Gene Aliases
Lplunc3, Rya3
Chromosome Location
Chr.3: 149401890 - 149416506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109208.1 NP_001102678.1 1 - 2 128 84

View Full Product Details