Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfatc2ip
Entrez Gene ID
308983
Gene Name
nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2 interacting protein
Gene Aliases
-
Chromosome Location
Chr.1: 197785966 - 197800943 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007692.1 NP_001007693.1 6 - 7 1096 74
GenBank mRNA BC079050.1 - 6 - 7 1096 74

View Full Product Details