Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ovch2
Entrez Gene ID
308919
Gene Name
ovochymase 2
Gene Aliases
Ovch2-ps1, RGD1564636
Chromosome Location
Chr.1: 172009585 - 172030164 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748959.3 XP_003749007.1 6 - 7 710 96

View Full Product Details