The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133564.1 | NP_598248.1 | 7 - 8 | 1232 | 57 |
XM_006235674.2 | XP_006235736.1 | 8 - 9 | 5075 | 57 | |
GenBank mRNA | AJ302054.1 | - | 7 - 8 | 1232 | 57 |