Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a31
Entrez Gene ID
689108
Gene Name
solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31
Gene Aliases
-
Chromosome Location
Chr.2: 127538324 - 127554276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001069558.6 XP_001069558.2 7 - 8 1218 113

View Full Product Details