The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039342.2 | NP_001034431.2 | 9 - 10 | 1087 | 64 |
XM_006227796.2 | XP_006227858.1 | 8 - 9 | 1099 | 64 | |
XM_006227797.2 | XP_006227859.1 | 8 - 9 | 1048 | 64 | |
GenBank mRNA | AY886527.1 | - | 9 - 10 | 1177 | 64 |