Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc3b
Entrez Gene ID
305705
Gene Name
leucine rich repeat containing 3B
Gene Aliases
RGD1311897
Chromosome Location
Chr.15: 11222960 - 11299563 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728176.4 XP_002728222.1 1 - 2 588 73

View Full Product Details