Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atxn7
Entrez Gene ID
361015
Gene Name
ataxin 7
Gene Aliases
RGD1562692
Chromosome Location
Chr.15: 12421533 - 12569650 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006251728.2 XP_006251790.1 8 - 9 1245 99
XM_008770600.1 XP_008768822.1 8 - 9 1245 99

View Full Product Details