Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc18
Entrez Gene ID
306278
Gene Name
leucine rich repeat containing 18
Gene Aliases
RGD1311738
Chromosome Location
Chr.16: 9235512 - 9259461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013999.1 NP_001014021.1 2 - 3 183 72
XM_008771097.1 XP_008769319.1 2 - 3 559 72
GenBank mRNA BC081908.1 - 2 - 3 183 72

View Full Product Details