Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp5b
Entrez Gene ID
171374
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases
-
Chromosome Location
Chr.7: 2504708 - 2511748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134364.1 NP_599191.1 3 - 4 502 95
GenBank mRNA BC099743.1 - 3 - 4 485 95
FQ216116.1 - 1 - 2 82 95
FQ230301.1 - 1 - 2 121 95
M19044.1 - 2 - 3 318 95

View Full Product Details