Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp39a1
Entrez Gene ID
301264
Gene Name
cytochrome P450, family 39, subfamily a, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.9: 19804079 - 19880346 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106893.1 NP_001100363.1 7 - 8 1247 129
XM_006244604.2 XP_006244666.1 7 - 8 1247 129
XM_006244605.1 XP_006244667.1 7 - 8 1247 129
XM_008766854.1 XP_008765076.1 7 - 8 1130 129
XM_008766855.1 XP_008765077.1 7 - 8 1128 129

View Full Product Details