Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il17a
Entrez Gene ID
301289
Gene Name
interleukin 17A
Gene Aliases
CTLA-8, IL-17, IL-17A, Il17
Chromosome Location
Chr.9: 26841299 - 26844786 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106897.1 NP_001100367.1 2 - 3 300 78

View Full Product Details