The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106874.1 | NP_001100344.1 | 4 - 5 | 506 | 65 |
XM_006244305.1 | XP_006244367.1 | 4 - 5 | 555 | 65 | |
XM_008766754.1 | XP_008764976.1 | 4 - 5 | 616 | 65 | |
GenBank mRNA | FM165287.1 | - | 4 - 5 | 443 | 65 |