Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcn12
Entrez Gene ID
680602
Gene Name
lipocalin 12
Gene Aliases
-
Chromosome Location
Chr.3: 2716547 - 2719135 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128138.1 NP_001121610.1 1 - 2 330 65
GenBank mRNA DQ537492.1 - 2 - 3 296 65

View Full Product Details