Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Naif1
Entrez Gene ID
687739
Gene Name
nuclear apoptosis inducing factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 11476328 - 11482629 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749447.3 XP_003749495.1 1 - 2 1369 71

View Full Product Details