The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107835.2 | NP_001101305.1 | 6 - 7 | 1222 | 75 |
XM_006233976.1 | XP_006234038.1 | 6 - 7 | 1356 | 75 | |
XM_006233977.2 | XP_006234039.1 | 7 - 8 | 1272 | 75 | |
XM_006233980.2 | XP_006234042.1 | 7 - 8 | 1180 | 75 | |
XM_008761750.1 | XP_008759972.1 | 7 - 8 | 2831 | 75 | |
XM_008761751.1 | XP_008759973.1 | 8 - 9 | 1437 | 75 | |
XM_008761752.1 | XP_008759974.1 | 6 - 7 | 1158 | 75 | |
GenBank mRNA | BC166990.1 | - | 6 - 7 | 1222 | 75 |