Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2d3c
Entrez Gene ID
362111
Gene Name
SH2 domain containing 3C
Gene Aliases
-
Chromosome Location
Chr.3: 11756427 - 11793549 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108579.1 NP_001102049.1 4 - 5 755 90
XM_008761759.1 XP_008759981.1 3 - 4 1622 90
XM_008761760.1 XP_008759982.1 2 - 3 1631 90
GenBank mRNA BC158641.1 - 3 - 4 264 90

View Full Product Details