Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde6a
Entrez Gene ID
307401
Gene Name
phosphodiesterase 6A, cGMP-specific, rod, alpha
Gene Aliases
-
Chromosome Location
Chr.18: 56544652 - 56617480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107386.1 NP_001100856.1 4 - 5 962 89

View Full Product Details