Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a20
Entrez Gene ID
293686
Gene Name
solute carrier family 22, member 20
Gene Aliases
Gm963
Chromosome Location
Chr.1: 221327608 - 221343965 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106327.1 NP_001099797.1 1 - 2 378 60
XM_006230750.2 XP_006230812.1 1 - 2 454 60

View Full Product Details