Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Drap1
Entrez Gene ID
293674
Gene Name
Dr1 associated protein 1 (negative cofactor 2 alpha)
Gene Aliases
-
Chromosome Location
Chr.1: 220803783 - 220806476 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077668.2 NP_001071136.1 4 - 5 532 128
GenBank mRNA BC127524.1 - 4 - 5 533 128

View Full Product Details