Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc41a2
Entrez Gene ID
362861
Gene Name
solute carrier family 41 (magnesium transporter), member 2
Gene Aliases
-
Chromosome Location
Chr.7: 26522314 - 26607861 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108742.1 NP_001102212.1 7 - 8 1486 84

View Full Product Details