The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135582.1 | NP_001129054.1 | 3 - 4 | 236 | 64 |
GenBank mRNA | FQ219839.1 | - | 2 - 3 | 205 | 64 |
FQ220026.1 | - | 2 - 3 | 205 | 64 | |
FQ224899.1 | - | 2 - 3 | 205 | 64 |