Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhs
Entrez Gene ID
317494
Gene Name
Nance-Horan syndrome (congenital cataracts and dental anomalies)
Gene Aliases
RGD1561818
Chromosome Location
Chr.X: 34313566 - 34675912 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773173.1 XP_008771395.1 4 - 5 1687 84

View Full Product Details