Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpped1
Entrez Gene ID
302890
Gene Name
calcineurin-like phosphoesterase domain containing 1
Gene Aliases
RGD1306568
Chromosome Location
Chr.10: 3655038 - 3774887 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013963.1 NP_001013985.1 2 - 3 342 61
GenBank mRNA BC081991.1 - 2 - 3 342 61

View Full Product Details