Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcdh18
Entrez Gene ID
295027
Gene Name
protocadherin 18
Gene Aliases
-
Chromosome Location
Chr.2: 138819957 - 138833930 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232322.2 XP_006232384.1 1 - 2 2972 86

View Full Product Details