Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc1a7
Entrez Gene ID
366432
Gene Name
solute carrier family 1 (glutamate transporter), member 7
Gene Aliases
-
Chromosome Location
Chr.5: 127571114 - 127616033 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108973.1 NP_001102443.1 5 - 6 941 101
XM_008763892.1 XP_008762114.1 5 - 6 870 101
GenBank mRNA JF422064.1 - 5 - 6 743 101
JF422065.1 - 4 - 5 527 101
JF422066.1 - 5 - 6 743 101
JF422067.1 - 5 - 6 743 101
JF422068.1 - 5 - 6 743 101

View Full Product Details