Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaaf1
Entrez Gene ID
361419
Gene Name
dynein, axonemal, assembly factor 1
Gene Aliases
Lrrc50, RGD1310542
Chromosome Location
Chr.19: 52225582 - 52245930 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014154.1 NP_001014176.1 6 - 7 906 77
GenBank mRNA BC079038.1 - 6 - 7 906 77

View Full Product Details