Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod4
Entrez Gene ID
295261
Gene Name
tropomodulin 4
Gene Aliases
-
Chromosome Location
Chr.2: 196295069 - 196299896 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106449.2 NP_001099919.1 1 - 2 127 125
GenBank mRNA FN800735.1 - 1 - 2 85 125
FQ214903.1 - 1 - 2 64 125
FQ216125.1 - 1 - 2 53 125
FQ217203.1 - 1 - 2 51 125

View Full Product Details