Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmod4
Entrez Gene ID
295261
Gene Name
tropomodulin 4
Gene Aliases
-
Chromosome Location
Chr.2: 196295069 - 196299896 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106449.2 NP_001099919.1 6 - 7 789 61
NM_001270650.1 NP_001257579.1 7 - 8 863 61
NM_001270651.1 NP_001257580.1 7 - 8 836 61
NM_001270652.1 NP_001257581.1 6 - 7 766 61
NM_001270653.1 NP_001257582.1 6 - 7 793 61
XM_006232855.2 XP_006232917.1 7 - 8 858 61
XM_008761282.1 XP_008759504.1 6 - 7 824 61
GenBank mRNA FM111895.1 - 3 - 4 245 61
FQ214903.1 - 6 - 7 726 61
FQ216236.1 - 4 - 5 460 61
FQ216249.1 - 7 - 8 786 61
FQ216534.1 - 6 - 7 692 61
FQ217203.1 - 6 - 7 713 61
FQ224853.1 - 6 - 7 714 61

View Full Product Details