Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmod4
Entrez Gene ID
295261
Gene Name
tropomodulin 4
Gene Aliases
-
Chromosome Location
Chr.2: 196295069 - 196299896 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106449.2 NP_001099919.1 9 - 10 1182 71
NM_001270650.1 NP_001257579.1 10 - 11 1256 71
NM_001270651.1 NP_001257580.1 10 - 11 1229 71
NM_001270652.1 NP_001257581.1 9 - 10 1159 71
NM_001270653.1 NP_001257582.1 9 - 10 1186 71
XM_006232855.2 XP_006232917.1 10 - 11 1251 71
XM_008761282.1 XP_008759504.1 9 - 10 1217 71
GenBank mRNA FQ214903.1 - 9 - 10 1119 71
FQ215988.1 - 10 - 11 1185 71
FQ216534.1 - 9 - 10 1085 71
FQ217203.1 - 9 - 10 1106 71
FQ224853.1 - 9 - 10 1107 71

View Full Product Details