Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dleu7
Entrez Gene ID
290308
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
RGD1305071
Chromosome Location
Chr.15: 45360503 - 45376807 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106043.1 NP_001099513.1 2 - 3 833 99

View Full Product Details