Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl6
Entrez Gene ID
363760
Gene Name
ADP-ribosylation factor-like 6
Gene Aliases
-
Chromosome Location
Chr.11: 42858467 - 42884324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108842.1 NP_001102312.1 6 - 7 737 87
XM_006248191.2 XP_006248253.1 6 - 7 1012 87
XM_006248192.2 XP_006248254.1 6 - 7 862 87
XM_006248193.2 XP_006248255.1 6 - 7 1011 87
GenBank mRNA BC162029.1 - 6 - 7 858 87

View Full Product Details