Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dirc2
Entrez Gene ID
303902
Gene Name
disrupted in renal carcinoma 2
Gene Aliases
-
Chromosome Location
Chr.11: 68198709 - 68272766 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012017.1 NP_001012017.1 2 - 3 695 75
GenBank mRNA BC081960.1 - 2 - 3 695 75

View Full Product Details