Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Raver2
Entrez Gene ID
362551
Gene Name
ribonucleoprotein, PTB-binding 2
Gene Aliases
RGD1307610
Chromosome Location
Chr.5: 119903449 - 119982687 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191867.1 NP_001178796.1 1 - 2 287 120
XM_006238451.2 XP_006238513.1 1 - 2 345 120

View Full Product Details