Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
L1td1
Entrez Gene ID
685355
Gene Name
LINE-1 type transposase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 117348106 - 117361012 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001063462.4 XP_001063462.2 2 - 3 169 71

View Full Product Details