Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr87
Entrez Gene ID
687472
Gene Name
WD repeat domain 87
Gene Aliases
-
Chromosome Location
Chr.1: 84828703 - 84858821 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008759284.1 XP_008757506.1 5 - 6 3480 121

View Full Product Details