Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhdh
Entrez Gene ID
691002
Gene Name
dihydrodiol dehydrogenase (dimeric)
Gene Aliases
-
Chromosome Location
Chr.1: 101465952 - 101476928 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229208.2 XP_006229270.1 1 - 2 731 66
XM_008759458.1 XP_008757680.1 2 - 3 190 66

View Full Product Details