Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syngr4
Entrez Gene ID
292916
Gene Name
synaptogyrin 4
Gene Aliases
-
Chromosome Location
Chr.1: 101872541 - 101883963 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025644.1 NP_001020815.1 2 - 3 308 103
XM_006229034.2 XP_006229096.1 2 - 3 524 103
XM_008759339.1 XP_008757561.1 2 - 3 527 103
GenBank mRNA BC098938.1 - 2 - 3 308 103

View Full Product Details